Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000399.4 | NP_000390.2 | 2 | 2074 | ||
NM_001136177.2 | NP_001129649.1 | 3 | 1891 | |||
NM_001136178.1 | NP_001129650.1 | 4 | 1932 | |||
NM_001136179.2 | NP_001129651.1 | 3 | 1974 | |||
NM_001321037.1 | NP_001307966.1 | 3 | 1791 | |||
XM_011539427.1 | XP_011537729.1 | 3 | 1777 | |||
GenBank mRNA | AK091399.1 | - | 3 | 1909 | ||
BC035625.1 | - | 2 | 2118 | 5208710 | ||
J04076.1 | - | 2 | 1778 |