Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527533.2 | XP_011525835.1 | 8 | 1569 | ||
XM_011527535.2 | XP_011525837.1 | 8 | 1483 | |||
XM_011527536.2 | XP_011525838.1 | 8 | 1408 | |||
XM_011527537.2 | XP_011525839.1 | 8 | 1375 | |||
XM_011527538.2 | XP_011525840.1 | 6 | 1395 | |||
GenBank mRNA | AK300170.1 | - | 7 | 1328 | ||
AK300182.1 | - | 7 | 1425 | |||
BC035359.1 | - | 7 | 1349 | 5176194 | ||
BC140798.1 | - | 6 | 1138 | 9021666 | ||
D86358.1 | - | 8 | 1531 | |||
D86359.1 | - | 6 | 1355 | |||
U71382.1 | - | 8 | 1325 |