Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001264.4 | NP_001255.3 | 2 | 1600 | ||
GenBank mRNA | AF030130.1 | - | 2 | 1558 | ||
AK292636.1 | - | 2 | 1568 | |||
BC031993.1 | - | 2 | 1548 | 4747362 | ||
L20815.1 | - | 2 | 1557 |