Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319165.1 | NP_001306094.1 | 4 | 1007 | ||
NM_001319166.1 | NP_001306095.1 | 4 | 744 | |||
NM_001319167.1 | NP_001306096.1 | 4 | 1074 | |||
NM_001319168.1 | NP_001306097.1 | 4 | 965 | |||
NM_001319169.1 | NP_001306098.1 | 3 | 793 | |||
NM_001319170.1 | NP_001306099.1 | 4 | 1077 | |||
NM_001319171.1 | NP_001306100.1 | 4 | 1048 | |||
NM_001384.4 | NP_001375.2 | 4 | 1017 | |||
XM_017000502.1 | XP_016855991.1 | 4 | 1019 | |||
XM_017000503.1 | XP_016855992.1 | 4 | 953 | |||
XM_017000504.1 | XP_016855993.1 | 3 | 1258 | |||
XM_017000505.1 | XP_016855994.1 | 3 | 1190 | |||
XM_017000506.1 | XP_016855995.1 | 4 | 1061 | |||
GenBank mRNA | AK091303.1 | - | 4 | 936 | ||
AK297933.1 | - | 3 | 766 | |||
AK315506.1 | - | 4 | 1001 | |||
BC001389.2 | - | 4 | 976 | 3051455 | ||
BC003181.1 | - | 4 | 994 | 2958202 | ||
BC016956.1 | - | 4 | 976 | 3900854 | ||
BT007431.1 | - | 4 | 822 |