Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001405.3 | NP_001396.2 | 4 | 580 | ||
XM_017026449.1 | XP_016881938.1 | 5 | 2385 | |||
XM_017026450.1 | XP_016881939.1 | 5 | 1269 | |||
GenBank mRNA | AJ007292.1 | - | 4 | 580 |