Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001419.2 | NP_001410.2 | 6 | 1455 | ||
GenBank mRNA | AK301013.1 | - | 6 | 1457 | ||
BC003376.2 | - | 6 | 1418 | 2901220 | ||
BX647856.1 | - | 1 | 167 |