Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001424.5 | NP_001415.1 | 5 | 592 | ||
XM_006720864.3 | XP_006720927.1 | 5 | 586 | |||
GenBank mRNA | AK313134.1 | - | 5 | 508 | ||
AV657588.1 | - | 4 | 307 | |||
BC009687.1 | - | 5 | 483 | 3895094 | ||
BF981157.1 | - | 5 | 574 | |||
U52100.1 | - | 4 | 427 | |||
X94770.1 | - | 4 | 374 |