Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145949
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
17
1553
NM_001190442.1
NP_001177371.1
18
1425
NM_001284212.2
NP_001271141.1
15
1204
NM_001284213.2
NP_001271142.1
14
1111
NM_173060.3
NP_775083.1
16
1261
NR_104285.1
4
417
XM_006714696.3
XP_006714759.1
20
1567
XM_006714697.3
XP_006714760.1
19
1584
XM_006714698.3
XP_006714761.1
18
1545
XM_006714699.3
XP_006714762.1
19
1508
XM_006714700.3
XP_006714763.1
18
1527
XM_006714701.3
XP_006714764.1
18
1543
XM_006714702.3
XP_006714765.1
18
1419
XM_006714703.3
XP_006714766.1
17
1504
XM_006714704.3
XP_006714767.1
18
1452
XM_006714705.3
XP_006714768.1
17
1461
XM_006714706.3
XP_006714769.1
17
1379
XM_006714707.3
XP_006714770.1
17
1330
XM_006714708.3
XP_006714771.1
16
1290
XM_006714709.3
XP_006714772.1
16
1253
XM_006714710.3
XP_006714773.1
15
1208
XM_006714711.3
XP_006714774.1
15
1253
XM_006714712.3
XP_006714775.1
14
1200
XM_006714713.3
XP_006714776.1
4
417
XM_006714714.3
XP_006714777.1
4
417
XM_006714715.3
XP_006714778.1
5
1143
XM_011543654.2
XP_011541956.1
18
1503
XM_011543655.2
XP_011541957.1
18
1500
XM_011543656.2
XP_011541958.1
17
1447
XM_011543657.2
XP_011541959.1
17
1329
XM_011543658.2
XP_011541960.1
15
1182
XM_017009911.1
XP_016865400.1
18
1545
XM_017009912.1
XP_016865401.1
19
1556
XM_017009913.1
XP_016865402.1
19
1525
XM_017009914.1
XP_016865403.1
18
1527
XM_017009915.1
XP_016865404.1
19
1529
XM_017009916.1
XP_016865405.1
18
1543
XM_017009917.1
XP_016865406.1
18
1461
XM_017009918.1
XP_016865407.1
17
1488
XM_017009919.1
XP_016865408.1
17
1461
XM_017009920.1
XP_016865409.1
17
1371
XM_017009921.1
XP_016865410.1
17
1388
XM_017009922.1
XP_016865411.1
16
1422
XM_017009923.1
XP_016865412.1
16
1422
XM_017009924.1
XP_016865413.1
16
1404
XM_017009925.1
XP_016865414.1
16
1340
XM_017009926.1
XP_016865415.1
16
1340
XM_017009927.1
XP_016865416.1
17
1329
XM_017009928.1
XP_016865417.1
16
1328
XM_017009929.1
XP_016865418.1
16
1247
XM_017009930.1
XP_016865419.1
15
1214
XM_017009931.1
XP_016865420.1
15
1208
XM_017009932.1
XP_016865421.1
14
1271
GenBank mRNA
AB208816.1
-
18
1389
AF327443.1
-
13
1103
AK292289.1
-
4
417
AK292356.1
-
4
390
AK296919.1
-
18
1383
AK297879.1
-
16
1203
AK298541.1
-
17
1330
AK299402.1
-
17
1310
AK300624.1
-
14
1094
AK302206.1
-
5
417
AK303836.1
-
10
784
AK311585.1
-
5
595
AK311598.1
-
5
471
AK314290.1
-
15
1206
AK315146.1
-
15
1205
AK315673.1
-
4
365
AL832174.1
-
16
1246
AL832349.1
-
16
1249
AY189741.1
-
6
416
BC013579.1
-
17
1256
3878564
BC078149.1
-
12
839
BT009783.1
-
14
1038
BX648397.1
-
15
1155
D16217.1
-
17
1323
DB033043.1
-
4
365
U26724.2
-
8
686
U31345.1
-
6
433
U58996.2
-
4
396
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0