Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001818.3 | NP_001809.3 | 9 | 1020 | ||
GenBank mRNA | BC020744.1 | - | 9 | 1020 | 4734943 | |
D26125.1 | - | 9 | 983 | |||
M33375.1 | - | 9 | 985 | |||
S68287.1 | - | 9 | 994 |
Gene Symbol: AKR1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M33376.1 | - | 9 | 942 |