Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025201.3 | NP_001020372.2 | 13 | 2125 | ||
NM_001206602.1 | NP_001193531.1 | 7 | 1973 | |||
NM_001822.5 | NP_001813.1 | 13 | 2202 | |||
NR_038133.1 | 12 | 1727 | ||||
GenBank mRNA | AB209788.1 | - | 12 | 1712 | ||
AK055060.1 | - | 7 | 1904 | |||
AK289941.1 | - | 13 | 2132 | |||
BC011393.1 | - | 13 | 2174 | 3871858 | ||
DB548770.1 | - | 1 | 131 | |||
X51408.1 | - | 1933 | ||||
Z22641.1 | - | 13 | 1871 |