Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286055.1 | NP_001272984.1 | 3 | 553 | ||
NM_001286056.1 | NP_001272985.1 | 3 | 343 | |||
NM_001839.4 | NP_001830.1 | 3 | 553 | |||
XM_017000245.1 | XP_016855734.1 | 5 | 2965 | |||
GenBank mRNA | AK294141.1 | - | 3 | 304 | ||
AK298145.1 | - | 3 | 473 | |||
AK312370.1 | - | 3 | 440 | |||
AK316398.1 | - | 3 | 319 | |||
BC025372.2 | - | 3 | 437 | 3505668 | ||
BU149686.1 | - | 3 | 489 | |||
CR541848.1 | - | 3 | 224 | |||
DR157009.1 | - | 3 | 476 | |||
S80562.1 | - | 3 | 307 |