Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286056.1 | NP_001272985.1 | 4 | 424 | ||
NM_001839.4 | NP_001830.1 | 4 | 634 | |||
XM_017000245.1 | XP_016855734.1 | 6 | 3046 | |||
GenBank mRNA | AK294141.1 | - | 4 | 385 | ||
AK312370.1 | - | 4 | 521 | |||
AK316398.1 | - | 4 | 400 | |||
BC025372.2 | - | 4 | 518 | 3505668 | ||
BU149686.1 | - | 4 | 570 | |||
CR541848.1 | - | 4 | 305 | |||
DR157009.1 | - | 4 | 557 | |||
S80562.1 | - | 4 | 388 |