Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286055.1 | NP_001272984.1 | 6 | 1246 | ||
NM_001286056.1 | NP_001272985.1 | 7 | 1174 | |||
NM_001839.4 | NP_001830.1 | 7 | 1384 | |||
XM_017000245.1 | XP_016855734.1 | 9 | 3796 | |||
GenBank mRNA | AK294141.1 | - | 7 | 1135 | ||
AK298145.1 | - | 6 | 1166 | |||
AK316398.1 | - | 7 | 1150 | |||
BC025372.2 | - | 7 | 1268 | 3505668 | ||
S80562.1 | - | 7 | 1138 |