Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165928.3 | NP_001159400.2 | 6 | 1090 | ||
NM_001177634.2 | NP_001171105.1 | 6 | 1033 | |||
NM_001177635.2 | NP_001171106.1 | 4 | 622 | |||
NM_001177636.2 | NP_001171107.1 | 5 | 1011 | |||
NM_001177637.2 | NP_001171108.1 | 4 | 849 | |||
NM_001177638.2 | NP_001171109.1 | 4 | 880 | |||
NM_001177639.2 | NP_001171110.1 | 3 | 570 | |||
NM_001177640.2 | NP_001171111.1 | 4 | 786 | |||
NM_001177641.2 | NP_001171112.1 | 4 | 876 | |||
NM_001177642.2 | NP_001171113.1 | 4 | 781 | |||
NM_001177643.2 | NP_001171114.1 | 3 | 780 | |||
NM_001177644.2 | NP_001171115.1 | 3 | 869 | |||
NM_004393.5 | NP_004384.4 | 3 | 729 | |||
GenBank mRNA | AK291692.1 | - | 3 | 571 | ||
BC012740.2 | - | 3 | 664 | 4099382 | ||
BC014616.2 | - | 3 | 664 | 2964740 | ||
DA223570.1 | - | 4 | 507 | |||
L19711.1 | - | 3 | 705 |