Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001888.4 | NP_001879.1 | 10 | 1469 | ||
GenBank mRNA | AK290852.1 | - | 8 | 1188 | ||
BC018061.1 | - | 8 | 1187 | 4817634 | ||
BX648477.1 | - | 10 | 1385 | |||
L02950.1 | - | 8 | 1148 | |||
U85772.1 | - | 8 | 1143 |