Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017916.1 | NP_001017916.1 | 3 | 543 | ||
NM_001017917.1 | NP_001017917.1 | 3 | 376 | |||
NM_001915.3 | NP_001906.3 | 3 | 373 | |||
XM_005257091.1 | XP_005257148.1 | 3 | 318 | |||
XM_017024260.1 | XP_016879749.1 | 3 | 415 | |||
XM_017024261.1 | XP_016879750.1 | 2 | 256 | |||
XM_017024262.1 | XP_016879751.1 | 3 | 608 | |||
GenBank mRNA | AK095244.1 | - | 3 | 543 | ||
AK294403.1 | - | 3 | 304 | |||
AK294630.1 | - | 3 | 527 | |||
AK301541.1 | - | 3 | 303 | |||
AK303177.1 | - | 3 | 242 | |||
AK316606.1 | - | 3 | 318 | |||
BC000021.1 | - | 3 | 404 | 3509626 | ||
BC002976.1 | - | 3 | 330 | 3535771 | ||
BC091485.1 | - | 3 | 344 | 6188259 | ||
BF206825.1 | - | 378 | ||||
BM552101.1 | - | 3 | 373 | |||
BT007096.1 | - | 2 | 243 |