Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168319.1 | NP_001161791.1 | 2, 3 | 573 | ||
NM_001955.4 | NP_001946.3 | 2, 3 | 576 | |||
XM_011514330.2 | XP_011512632.1 | 3, 4 | 332 | |||
XM_011514331.2 | XP_011512633.1 | 4, 5 | 493 | |||
XM_011514332.2 | XP_011512634.1 | 3, 4 | 330 | |||
XM_017010331.1 | XP_016865820.1 | 3, 4 | 516 | |||
GenBank mRNA | AK226096.1 | - | 2, 3 | 491 | ||
AK291838.1 | - | 2, 3 | 513 | |||
AK313914.1 | - | 2, 3 | 508 | |||
BC009720.1 | - | 2, 3 | 445 | 3900704 | ||
BG540968.1 | - | 510 | ||||
BX439515.2 | - | 2, 3 | 536 | |||
CR541903.1 | - | 2, 3 | 242 | |||
S56805.1 | - | 2, 3 | 578 | |||
Y00749.1 | - | 2, 3 | 494 |