Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMTN SIRNA Assays ›
Gene Symbol:
DMTN
Gene Name:
dematin actin binding protein
Gene Aliases:
DMT, EPB49
Chromosome Location:
Chr.8: 22048955 - 22082527 on Build GRCh38
UniGene ID:
Hs.106124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146241
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114135.4
NP_001107607.1
8
830
NM_001114136.2
NP_001107608.1
8
786
NM_001114137.3
NP_001107609.1
8
960
NM_001114138.2
NP_001107610.1
8
947
NM_001114139.3
NP_001107611.1
7
844
NM_001302816.2
NP_001289745.1
8
1080
NM_001302817.2
NP_001289746.1
6
799
NM_001323378.1
NP_001310307.1
8
919
NM_001323380.1
NP_001310309.1
8
826
NM_001323381.1
NP_001310310.1
7
935
NM_001323382.1
NP_001310311.1
8
865
NM_001323383.1
NP_001310312.1
8
786
NM_001323384.1
NP_001310313.1
8
919
NM_001323385.1
NP_001310314.1
7
935
NM_001323387.1
NP_001310316.1
7
884
NM_001323388.1
NP_001310317.1
6
666
NM_001323389.1
NP_001310318.1
7
874
NM_001323390.1
NP_001310319.1
7
741
NM_001323391.1
NP_001310320.1
7
844
NM_001323392.1
NP_001310321.1
7
1005
NM_001323393.1
NP_001310322.1
7
711
NM_001323394.1
NP_001310323.1
7
872
NM_001323395.1
NP_001310324.1
7
751
NM_001323396.1
NP_001310325.1
7
755
NM_001323397.1
NP_001310326.1
7
923
NM_001323398.1
NP_001310327.1
6
799
NM_001323399.1
NP_001310328.1
6
666
NM_001323400.1
NP_001310329.1
5
815
NM_001323401.1
NP_001310330.1
6
839
NM_001978.4
NP_001969.2
8
1080
XM_005273432.1
XP_005273489.1
8
812
XM_005273433.1
XP_005273490.1
8
744
XM_005273442.1
XP_005273499.1
7
596
XM_011544435.1
XP_011542737.1
7
1005
XM_017013189.1
XP_016868678.1
8
753
XM_017013190.1
XP_016868679.1
7
1005
XM_017013191.1
XP_016868680.1
7
759
XM_017013192.1
XP_016868681.1
7
640
XM_017013193.1
XP_016868682.1
6
772
XM_017013194.1
XP_016868683.1
6
639
XM_017013195.1
XP_016868684.1
7
1005
XM_017013196.1
XP_016868685.1
7
608
XM_017013197.1
XP_016868686.1
7
839
XM_017013198.1
XP_016868687.1
7
681
GenBank mRNA
AB209718.1
-
4
306
AK055842.1
-
8
744
AK091581.1
-
7
759
AK289650.1
-
8
712
AK295452.1
-
6
685
AK308807.1
-
6
979
BC006318.2
-
8
829
4125804
BC017445.1
-
8
829
3162732
BC052805.1
-
8
870
6269658
BT007396.1
-
7
557
L19713.1
-
8
1007
U28389.1
-
8
1007
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
transmembrane transport
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
protein binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0