Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320747.1 | NP_001307676.1 | 22 | 6473 | ||
NM_001988.3 | NP_001979.2 | 22 | 6407 | |||
GenBank mRNA | BX114092.1 | - | 1 | 194 | ||
U53786.1 | - | 22 | 6251 |