Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144663.1 | NP_001138135.1 | 4 | 391 | ||
NM_004063.3 | NP_004054.3 | 4 | 390 | |||
XM_011516790.1 | XP_011515092.1 | 4 | 389 | |||
GenBank mRNA | AK301001.1 | - | 4 | 358 | ||
AK312873.1 | - | 4 | 391 | |||
BC112013.1 | - | 4 | 363 | 8327481 | ||
BC113464.1 | - | 4 | 351 | 8322516 | ||
DB009107.1 | - | 4 | 389 | |||
U07969.1 | - | 4 | 352 | |||
X83228.1 | - | 4 | 357 |