Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039936.2 | NP_001035025.1 | 7 | 1752 | ||
NM_001293069.1 | NP_001279998.1 | 14 | 2023 | |||
NM_001293070.1 | NP_001279999.1 | 14 | 2237 | |||
NM_001293071.1 | NP_001280000.1 | 12 | 2142 | |||
NM_001293072.1 | NP_001280001.1 | 13 | 1955 | |||
NM_001293073.1 | NP_001280002.1 | 7 | 1617 | |||
NM_001293075.1 | NP_001280004.1 | 6 | 1479 | |||
NM_001293076.1 | NP_001280005.1 | 6 | 1578 | |||
NM_001293077.1 | NP_001280006.1 | 5 | 1536 | |||
NM_001293078.1 | NP_001280007.1 | 5 | 1440 | |||
NM_001293079.1 | NP_001280008.1 | 5 | 1508 | |||
NM_001293080.1 | NP_001280009.1 | 6 | 1614 | |||
NM_001293081.1 | NP_001280010.1 | 7 | 1267 | |||
NM_004067.3 | NP_004058.1 | 13 | 2198 | |||
NR_120524.1 | 4 | 1288 | ||||
NR_120525.1 | 5 | 1358 | ||||
XM_011515105.2 | XP_011513407.1 | 16 | 2078 | |||
XM_011515106.2 | XP_011513408.1 | 15 | 2039 | |||
XM_011515107.2 | XP_011513409.1 | 15 | 2821 | |||
XM_017011721.1 | XP_016867210.1 | 17 | 2096 | |||
XM_017011722.1 | XP_016867211.1 | 16 | 2839 | |||
GenBank mRNA | AK026415.1 | - | 7 | 1752 | ||
AK126784.1 | - | 2 | 2956 | |||
AK294216.1 | - | 7 | 1267 | |||
AK294271.1 | - | 14 | 2024 | |||
AK316030.1 | - | 7 | 1276 | |||
U28926.1 | - | 13 | 1990 |