Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004344.1 | NP_004335.1 | 5 | 561 | ||
GenBank mRNA | BC005334.1 | - | 5 | 561 | 3961448 | |
BC013873.2 | - | 5 | 559 | 3836808 | ||
X72964.1 | - | 5 | 561 |