Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127453.1 | NP_001120925.1 | 4 | 555 | ||
NM_001127454.1 | NP_001120926.1 | 3 | 570 | |||
NM_004403.2 | NP_004394.1 | 4 | 800 | |||
XM_017011802.1 | XP_016867291.1 | 3 | 326 | |||
GenBank mRNA | AF007790.2 | - | 3 | 455 | ||
AF073308.1 | - | 4 | 524 | |||
AK094714.1 | - | 6 | 1128 | |||
AK290775.1 | - | 4 | 531 | |||
AK314402.1 | - | 4 | 540 | |||
BC099911.1 | - | 4 | 516 | 5287362 | ||
BC125065.1 | - | 4 | 522 | 40117291 | ||
BC125066.1 | - | 3 | 292 |