Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004629.1 | NP_004620.1 | 12, 13 | 2136 | ||
GenBank mRNA | AJ007669.1 | - | 12, 13 | 2136 | ||
AK293427.1 | - | 12, 13 | 1674 | |||
AK311348.1 | - | 11, 12 | 1610 | |||
AK312987.1 | - | 12, 13 | 2061 | |||
BC000032.2 | - | 12, 13 | 1782 | 3507258 | ||
BC011623.2 | - | 12, 13 | 1909 | 4053144 | ||
BT009813.1 | - | 12, 13 | 1644 | |||
KU159422.1 | - | 12, 13 | 2149 | |||
KU178053.1 | - | 12, 13 | 1644 | |||
KU178054.1 | - | 12, 13 | 1626 | |||
U70310.1 | - | 12, 13 | 1778 |