Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDH18 SIRNA Assays ›
Gene Symbol:
CDH18
Gene Name:
cadherin 18
Gene Aliases:
CDH14, CDH14L, CDH24
Chromosome Location:
Chr.5: 19472930 - 20575902 on Build GRCh38
UniGene ID:
Hs.317632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146544
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDH18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001167667.1
NP_001161139.1
4
899
NM_001291956.1
NP_001278885.1
6
1356
NM_001291957.1
NP_001278886.1
4
877
NM_004934.3
NP_004925.1
4
899
XM_005248228.3
XP_005248285.1
4
1108
XM_006714435.3
XP_006714498.1
3
715
XM_011513928.2
XP_011512230.1
2
513
XM_011513930.2
XP_011512232.1
4
986
XM_017008924.1
XP_016864413.1
5
1227
XM_017008925.1
XP_016864414.1
5
1223
XM_017008926.1
XP_016864415.1
5
1084
XM_017008927.1
XP_016864416.1
5
1013
XM_017008928.1
XP_016864417.1
6
1271
XM_017008929.1
XP_016864418.1
3
751
XM_017008930.1
XP_016864419.1
3
642
XM_017008931.1
XP_016864420.1
4
895
XM_017008932.1
XP_016864421.1
5
946
XM_017008933.1
XP_016864422.1
4
986
XM_017008934.1
XP_016864423.1
4
1124
XM_017008935.1
XP_016864424.1
5
1227
XM_017008936.1
XP_016864425.1
4
1108
XM_017008937.1
XP_016864426.1
4
867
XM_017008938.1
XP_016864427.1
4
831
XM_017008939.1
XP_016864428.1
5
1204
XM_017008940.1
XP_016864429.1
4
1092
XM_017008941.1
XP_016864430.1
4
851
GenBank mRNA
AK289547.1
-
6
1354
AK295087.1
-
4
840
AK308745.1
-
4
877
BC031051.1
-
4
855
5278717
U59325.1
-
3
678
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
adherens junction organization
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0