Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNB SIRNA Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
UniGene ID:
Hs.307720
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146769
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256303.1
NP_001243232.1
3, 4
410
NM_001256304.1
NP_001243233.1
2, 3
147
NM_001256308.2
NP_001243237.1
2, 3
281
NM_001320932.1
NP_001307861.1
3, 4
410
NM_001320933.1
NP_001307862.1
3, 4
410
NM_001320934.1
NP_001307863.1
3, 4
410
NM_001320935.1
NP_001307864.1
3, 4
410
NM_001320936.1
NP_001307865.1
3, 4
410
NM_001320937.1
NP_001307866.1
3, 4
410
NM_021907.4
NP_068707.1
3, 4
410
NM_033147.3
NP_149159.2
3, 4
410
NM_033148.3
NP_149160.1
3, 4
410
NM_183360.2
NP_899204.1
3, 4
410
NM_183361.2
NP_899205.1
3, 4
410
XM_005264182.1
XP_005264239.1
3, 4
327
XM_011532671.1
XP_011530973.1
3, 4
327
XM_011532672.2
XP_011530974.1
3, 4
327
XM_011532673.1
XP_011530975.1
3, 4
327
XM_011532674.1
XP_011530976.1
3, 4
327
XM_011532675.1
XP_011530977.1
3, 4
327
XM_011532676.1
XP_011530978.1
3, 4
327
XM_011532677.2
XP_011530979.1
2, 3
783
XM_011532679.1
XP_011530981.1
3, 4
327
XM_011532680.2
XP_011530982.1
3, 4
334
XM_011532681.2
XP_011530983.1
3, 4
327
XM_017003529.1
XP_016859018.1
3, 4
327
XM_017003530.1
XP_016859019.1
3, 4
327
XM_017003531.1
XP_016859020.1
3, 4
327
XM_017003532.1
XP_016859021.1
3, 4
327
XM_017003533.1
XP_016859022.1
3, 4
327
XM_017003534.1
XP_016859023.1
3, 4
327
XM_017003535.1
XP_016859024.1
3, 4
327
XM_017003536.1
XP_016859025.1
3, 4
327
XM_017003538.1
XP_016859027.1
3, 4
259
XM_017003539.1
XP_016859028.1
3, 4
270
XM_017003540.1
XP_016859029.1
3, 4
327
XM_017003541.1
XP_016859030.1
3, 4
311
XM_017003542.1
XP_016859031.1
3, 4
334
XM_017003543.1
XP_016859032.1
3, 4
314
XM_017003544.1
XP_016859033.1
3, 4
334
XM_017003545.1
XP_016859034.1
3, 4
314
GenBank mRNA
AF022728.1
-
3, 4
308
AK094494.1
-
3, 4
273
AK225724.1
-
3, 4
313
AK294174.1
-
3, 4
315
AK294357.1
-
3, 4
177
AK300019.1
-
3, 4
287
AK301386.1
-
2, 3
283
AL555818.3
-
3, 4
324
BC016655.1
-
3, 4
303
3868207
BC049366.1
-
3, 4
401
4817552
BT009805.1
-
2, 3
147
BU846095.1
-
1, 2
88
DB460644.1
-
3, 4
410
DQ160290.1
-
2, 3
147
Y12712.1
-
3, 4
330
Y15718.1
-
2, 3
147
Y15722.1
-
2, 3
147
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0