Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001224.4 | NP_001215.1 | 12 | 1821 | ||
NM_032982.3 | NP_116764.2 | 11 | 2006 | |||
NM_032983.3 | NP_116765.2 | 10 | 1788 | |||
GenBank mRNA | AB209640.1 | - | 5 | 4678 | ||
BC002427.2 | - | 11 | 1778 | 3346279 | ||
BX537669.1 | - | 1 | 3506 | |||
U13022.1 | - | 12 | 1822 |