Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018115.2 | NP_001018125.1 | 38, 39 | 3977 | ||
NM_001319984.1 | NP_001306913.1 | 38, 39 | 4049 | |||
NM_033084.4 | NP_149075.2 | 38, 39 | 3977 | |||
GenBank mRNA | AF230336.1 | - | 38, 39 | 3920 | ||
AF340183.1 | - | 38, 39 | 3920 | |||
AK074406.1 | - | 11, 12 | 1175 | |||
AL832427.1 | - | 21, 22 | 2262 |