Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025160.2 | NP_001020331.1 | 17 | 2544 | ||
NM_001784.4 | NP_001775.2 | 16 | 2412 | |||
NM_078481.3 | NP_510966.1 | 18 | 2691 | |||
XM_011528451.2 | XP_011526753.1 | 18 | 2643 | |||
XM_017027547.1 | XP_016883036.1 | 19 | 2555 | |||
GenBank mRNA | AK097264.1 | - | 16 | 2007 | ||
AK225655.1 | - | 17 | 2253 | |||
AK225677.1 | - | 12 | 1230 | |||
AK292159.1 | - | 16 | 2399 | |||
AK304553.1 | - | 14 | 1851 | |||
AK308368.1 | - | 16 | 2218 | |||
AK314697.1 | - | 18 | 2391 | |||
AX775807.1 | - | 1413 | ||||
AX775809.1 | - | 1413 | ||||
AX775811.1 | - | 16 | 2120 | |||
AX775813.1 | - | 18 | 2359 | |||
BC026690.2 | - | 16 | 2108 | 3847171 | ||
U76764.1 | - | 18 | 2359 | |||
X84700.1 | - | 16 | 2102 |
Set Membership: |
G-protein coupled receptor |