Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000065.3 | NP_000056.2 | 5 | 770 | ||
NM_001115131.2 | NP_001108603.2 | 5 | 797 | |||
XM_005248357.2 | XP_005248414.1 | 5 | 845 | |||
XM_006714496.3 | XP_006714559.1 | 5 | 845 | |||
XM_011514114.2 | XP_011512416.1 | 5 | 845 | |||
XM_011514115.2 | XP_011512417.1 | 5 | 766 | |||
XM_011514116.2 | XP_011512418.1 | 6 | 882 | |||
XM_011514117.2 | XP_011512419.1 | 6 | 924 | |||
XM_011514118.2 | XP_011512420.1 | 5 | 662 | |||
XM_011514119.2 | XP_011512421.1 | 5 | 845 | |||
XM_017009818.1 | XP_016865307.1 | 6 | 905 | |||
GenBank mRNA | AB126592.1 | - | 4 | 532 | ||
AK292509.1 | - | 5 | 668 | |||
AK313673.1 | - | 5 | 689 | |||
BC035723.1 | - | 5 | 797 | 5591446 | ||
J04506.1 | - | 5 | 598 | |||
J05024.1 | - | 5 | 589 | |||
J05064.1 | - | 5 | 687 |