Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000668.5 | NP_000659.2 | 9 | 2445 | ||
NM_001286650.1 | NP_001273579.1 | 10 | 2567 | |||
GenBank mRNA | AF153821.1 | - | 9 | 2402 | ||
D00137.1 | - | 9 | 2354 | |||
FJ224285.1 | - | 9 | 2404 | |||
M21692.1 | - | 9 | 2365 | |||
X03350.1 | - | 9 | 2404 |