Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104.3 | NP_001095.2 | 6 | 714 | ||
NM_001258371.2 | NP_001245300.2 | 6 | 862 | |||
NR_047663.2 | 6 | 714 | ||||
GenBank mRNA | AK125851.1 | - | 6 | 1248 | ||
AK303044.1 | - | 6 | 862 | |||
AK313556.1 | - | 6 | 652 | |||
BC099647.3 | - | 6 | 635 | 40008220 | ||
BC099649.3 | - | 6 | 635 | 40008223 | ||
M86407.1 | - | 6 | 635 |