Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030007.1 | NP_001025178.1 | 15 | 1619 | ||
NM_001128.5 | NP_001119.3 | 14 | 1610 | |||
GenBank mRNA | AB015317.1 | - | 14 | 1315 | ||
AK128078.1 | - | 17 | 2165 | |||
AK225906.1 | - | 15 | 1378 | |||
AK299630.1 | - | 16 | 1553 | |||
AK314484.1 | - | 14 | 1615 | |||
BC036283.1 | - | 15 | 1584 | 4158263 | ||
Y12226.1 | - | 15 | 1333 |