Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129.4 | NP_001120.3 | 11 | 1591 | ||
XM_011515162.1 | XP_011513464.1 | 10 | 1554 | |||
GenBank mRNA | AB209744.1 | - | 11 | 1545 | ||
AF053944.1 | - | 11 | 1425 | |||
BC038588.1 | - | 11 | 1591 | 5764769 | ||
D86479.1 | - | 5 | 347 |