Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204.6 | NP_001195.2 | 13 | 5237 | ||
XM_011511687.1 | XP_011509989.1 | 13 | 5251 | |||
GenBank mRNA | BC032011.1 | - | 1 | 126 | ||
BG530017.1 | - | 1 | 292 |