Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204406.1 | NP_001191335.1 | 6 | 1052 | ||
NM_001629.3 | NP_001620.2 | 5 | 731 | |||
XM_017020522.1 | XP_016876011.1 | 4 | 582 | |||
GenBank mRNA | BC018538.1 | - | 5 | 684 | 4342203 |