Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142276.1 | NP_001135748.1 | 3 | 528 | ||
NM_001142277.1 | NP_001135749.1 | 3 | 528 | |||
NM_001142278.1 | NP_001135750.1 | 3 | 528 | |||
NM_001243299.1 | NP_001230228.1 | 3 | 446 | |||
NM_001642.2 | NP_001633.1 | 3 | 528 | |||
GenBank mRNA | AF168956.1 | - | 3 | 374 | ||
AK128162.1 | - | 3 | 445 | |||
AK225283.1 | - | 3 | 455 | |||
AK296765.1 | - | 3 | 455 | |||
AK304736.1 | - | 3 | 437 | |||
BC000373.2 | - | 3 | 428 | 2820109 | ||
BC004371.1 | - | 3 | 433 | 3640033 | ||
DC336210.1 | - | 3 | 528 | |||
L09209.1 | - | 3 | 443 | |||
L19597.1 | - | 3 | 439 | |||
L23114.1 | - | 2 | 259 | |||
L27631.1 | - | 3 | 444 | |||
S60099.1 | - | 3 | 443 | |||
Z22572.1 | - | 3 | 438 |