Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001661.3 | NP_001652.2 | 2 | 1481 | ||
XM_011524782.2 | XP_011523084.1 | 2 | 1409 | |||
GenBank mRNA | BU729196.1 | - | 1 | 251 |