Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BRDT SIRNA Assays ›
Gene Symbol:
BRDT
Gene Name:
bromodomain testis associated
Gene Aliases:
BRD6, CT9
Chromosome Location:
Chr.1: 91949371 - 92014428 on Build GRCh38
UniGene ID:
Hs.482520
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147246
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BRDT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242805.2
NP_001229734.2
9, 10
1710
NM_001242806.2
NP_001229735.2
8, 9
1653
NM_001242807.2
NP_001229736.2
7, 8
1309
NM_001242808.2
NP_001229737.2
7, 8
1503
NM_001242810.2
NP_001229739.2
8, 9
1997
NM_001726.4
NP_001717.3
8, 9
1436
NM_207189.3
NP_997072.2
8, 9
1641
XM_006710853.3
XP_006710916.1
9, 10
2226
XM_006710854.3
XP_006710917.1
10, 11
2071
XM_006710855.3
XP_006710918.1
10, 11
2076
XM_006710856.3
XP_006710919.1
10, 11
2050
XM_006710857.3
XP_006710920.1
10, 11
2040
XM_011542032.2
XP_011540334.1
10, 11
1560
XM_011542033.2
XP_011540335.1
10, 11
1534
XM_011542034.2
XP_011540336.1
10, 11
1550
XM_011542035.2
XP_011540337.1
10, 11
1524
XM_011542036.2
XP_011540338.1
8, 9
1439
GenBank mRNA
AF019085.1
-
8, 9
1399
AK302758.1
-
7, 8
1245
AK303008.1
-
8, 9
1401
AK316442.1
-
7, 8
1308
AY338951.1
-
9, 10
1710
BC005281.1
-
8, 9
1367
BC017582.1
-
8, 9
1442
BC047900.1
-
8, 9
1406
BC062700.1
-
8, 9
1401
BX648135.1
-
8, 9
1732
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone displacement
chromatin remodeling
transcription, DNA-templated
mRNA processing
male meiosis
male meiosis I
spermatogenesis
RNA splicing
covalent chromatin modification
cell differentiation
regulation of RNA splicing
positive regulation of transcription involved in meiotic cell cycle
transcription coactivator activity
histone binding
lysine-acetylated histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0