Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004436.2 | NP_004427.1 | 3 | 362 | ||
NM_207042.1 | NP_996925.1 | 4 | 410 | |||
NM_207043.1 | NP_996926.1 | 4 | 410 | |||
NM_207044.1 | NP_996927.1 | 3 | 362 | |||
NM_207045.1 | NP_996928.1 | 4 | 274 | |||
NM_207046.1 | NP_996929.1 | 3 | 226 | |||
NM_207047.1 | NP_996930.1 | 3 | 226 | |||
GenBank mRNA | AF067170.1 | - | 3 | 319 | ||
AF157509.1 | - | 3 | 316 | |||
AF157510.1 | - | 3 | 316 | |||
AK001981.1 | - | 3 | 336 | |||
AK290064.1 | - | 3 | 209 | |||
AL711949.1 | - | 3 | 365 | |||
BC000436.2 | - | 3 | 310 | 2820528 | ||
BC004461.1 | - | 3 | 310 | 2820528 | ||
BC068544.1 | - | 3 | 327 | 5273162 | ||
BE799496.1 | - | 3 | 246 | |||
BQ883543.1 | - | 3 | 324 | |||
BU858280.1 | - | 268 | ||||
CR457130.1 | - | 3 | 210 | |||
CR536578.1 | - | 3 | 210 | |||
CR749580.1 | - | 3 | 386 | |||
T78564.1 | - | 3 | 242 | |||
X99906.1 | - | 3 | 335 |