Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004446.2 | NP_004437.2 | 6 | 869 | ||
GenBank mRNA | AK296755.1 | - | 6 | 741 | ||
AK307534.1 | - | 4 | 443 | |||
AY493416.1 | - | 4 | 416 | |||
BC015494.1 | - | 6 | 657 | |||
BC034797.1 | - | 6 | 665 | |||
BC046156.1 | - | 6 | 869 | |||
BC058921.1 | - | 6 | 706 | |||
BC105983.1 | - | 6 | 667 | |||
BC126275.1 | - | 6 | 737 | 8991991 | ||
BC136465.1 | - | 6 | 637 | 9020454 | ||
CR933648.1 | - | 6 | 708 | |||
JQ080075.1 | - | 6 | 599 | |||
X07466.1 | - | 2 | 99 | |||
X54326.1 | - | 4 | 441 |