Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172415.1 | NP_001165886.1 | 4 | 776 | ||
NM_004323.5 | NP_004314.5 | 4 | 776 | |||
GenBank mRNA | AF022224.1 | - | 4 | 734 | ||
AF116273.1 | - | 4 | 704 | |||
AK093238.1 | - | 1 | 1435 | |||
AK096038.1 | - | 5 | 539 | |||
AK222749.1 | - | 4 | 723 | |||
AK316577.1 | - | 4 | 702 | |||
BC001936.1 | - | 4 | 737 | 2823774 | ||
BC014774.1 | - | 4 | 586 | |||
KU177932.1 | - | 4 | 476 | |||
KU177933.1 | - | 5 | 618 | |||
U46917.1 | - | 4 | 736 | |||
Z35491.1 | - | 4 | 754 |