Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004326.3 | NP_004317.2 | 7 | 1247 | ||
XM_005272971.4 | XP_005273028.1 | 7 | 1252 | |||
XM_005277417.3 | XP_005277474.1 | 7 | 1276 | |||
XM_005277418.3 | XP_005277475.1 | 7 | 1132 | |||
XM_017002026.1 | XP_016857515.1 | 7 | 1030 | |||
GenBank mRNA | BC116451.1 | - | 4 | 642 | 7961941 | |
Y13620.1 | - | 7 | 1335 |