Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006134.6 | NP_006125.2 | 7 | 1516 | ||
NR_040016.1 | 7 | 1516 | ||||
XM_011529746.1 | XP_011528048.1 | 7 | 1514 | |||
GenBank mRNA | AL355685.1 | - | 1 | 163 | ||
AY358634.1 | - | 7 | 1438 |