Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135934.1 | NP_001129406.1 | 7, 8 | 1021 | ||
NM_001135935.1 | NP_001129407.1 | 7, 8 | 1021 | |||
NM_001135936.1 | NP_001129408.1 | 7, 8 | 1021 | |||
NM_001286665.1 | NP_001273594.1 | 7, 8 | 1021 | |||
NM_001286666.1 | NP_001273595.1 | 7, 8 | 1021 | |||
NM_001286667.1 | NP_001273596.1 | 7, 8 | 1021 | |||
NM_006475.2 | NP_006466.2 | 7, 8 | 1021 | |||
XM_005266231.2 | XP_005266288.1 | 7, 8 | 1016 | |||
XM_005266232.2 | XP_005266289.1 | 7, 8 | 1016 | |||
XM_017020355.1 | XP_016875844.1 | 7, 8 | 1016 | |||
XM_017020356.1 | XP_016875845.1 | 7, 8 | 1016 | |||
GenBank mRNA | AK300261.1 | - | 7, 8 | 1135 | ||
AY140646.1 | - | 7, 8 | 914 | |||
AY918092.1 | - | 7, 8 | 914 | |||
BC106709.1 | - | 7, 8 | 914 | 40007999 | ||
BC106710.1 | - | 7, 8 | 914 | 40008000 | ||
BP335818.1 | - | 1, 2 | 57 | |||
BY795054.2 | - | 7, 8 | 928 | |||
D13665.1 | - | 7, 8 | 930 | |||
D13666.1 | - | 7, 8 | 914 | |||
EU262883.1 | - | 7, 8 | 903 | |||
EU262884.1 | - | 7, 8 | 903 | |||
EU262885.1 | - | 7, 8 | 903 | |||
EU262886.1 | - | 7, 8 | 903 | |||
GU354210.1 | - | 7, 8 | 914 |