Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015117.2 | NP_055932.2 | 12 | 3241 | ||
XM_006716536.3 | XP_006716599.2 | 14 | 3630 | |||
XM_017013248.1 | XP_016868737.1 | 13 | 4261 | |||
XM_017013249.1 | XP_016868738.1 | 13 | 3803 | |||
GenBank mRNA | BC038670.1 | - | 12 | 3224 | 5260824 | |
D63484.1 | - | 12 | 3201 |