Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015150.1 | NP_055965.1 | 10 | 2881 | ||
XM_005264985.1 | XP_005265042.1 | 10 | 2838 | |||
XM_005264986.2 | XP_005265043.1 | 10 | 2735 | |||
XM_006713069.1 | XP_006713132.1 | 10 | 2744 | |||
XM_011533529.1 | XP_011531831.1 | 12 | 3278 | |||
XM_011533530.1 | XP_011531832.1 | 10 | 2906 | |||
XM_011533531.1 | XP_011531833.1 | 11 | 3162 | |||
XM_017006007.1 | XP_016861496.1 | 9 | 2749 | |||
GenBank mRNA | AK074446.1 | - | 10 | 2688 | ||
AY237654.1 | - | 10 | 2872 | |||
BC006400.2 | - | 6 | 1809 | |||
BC014032.2 | - | 6 | 1809 | |||
BC032349.1 | - | 9 | 2554 | |||
BC051336.1 | - | 10 | 2658 | |||
BC069209.1 | - | 10 | 2658 | |||
D42043.1 | - | 10 | 2809 |