Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271949.1 | NP_001258878.1 | 7 | 1031 | ||
NM_001271950.1 | NP_001258879.1 | 6 | 1007 | |||
NM_015476.3 | NP_056291.2 | 7 | 1136 | |||
XM_017025699.1 | XP_016881188.1 | 8 | 1382 | |||
GenBank mRNA | AB073382.1 | - | 6 | 777 | ||
AK000543.1 | - | 7 | 709 | |||
AK295817.1 | - | 7 | 778 | |||
AL050225.1 | - | 6 | 595 | |||
AY598336.1 | - | 5 | 507 | |||
BC015178.1 | - | 7 | 897 | 3905315 | ||
BC087845.1 | - | 7 | 1115 | 30390077 |