Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZZZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308237.1 | NP_001295166.1 | 8 | 962 | ||
NM_015534.5 | NP_056349.1 | 9 | 2426 | |||
XM_005270725.3 | XP_005270782.1 | 9 | 2501 | |||
XM_005270726.3 | XP_005270783.1 | 8 | 2402 | |||
XM_005270727.3 | XP_005270784.1 | 9 | 2498 | |||
XM_005270729.4 | XP_005270786.1 | 7 | 912 | |||
XM_017000970.1 | XP_016856459.1 | 8 | 2399 | |||
XM_017000972.1 | XP_016856461.1 | 8 | 930 | |||
GenBank mRNA | AK074119.1 | - | 5 | 1472 | ||
BC035079.2 | - | 8 | 954 | |||
BC035397.1 | - | 5 | 1094 | |||
BC035818.2 | - | 5 | 2001 | 5554765 | ||
BX640658.1 | - | 5 | 514 | |||
BX640766.1 | - | 5 | 1700 | |||
BX641001.1 | - | 9 | 2427 | |||
BX641096.1 | - | 9 | 2412 |