Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114093.1 | NP_001107565.1 | 1 | 287 | ||
NM_015578.2 | NP_056393.2 | 1 | 287 | |||
XM_005258719.4 | XP_005258776.1 | 1 | 306 | |||
XM_005258720.4 | XP_005258777.1 | 1 | 306 | |||
XM_005258721.4 | XP_005258778.1 | 1 | 306 | |||
XM_011526708.2 | XP_011525010.1 | 1 | 306 | |||
XM_017026575.1 | XP_016882064.1 | 1 | 306 | |||
XM_017026576.1 | XP_016882065.1 | 1 | 306 | |||
XM_017026577.1 | XP_016882066.1 | 1 | 306 | |||
GenBank mRNA | AB069974.1 | - | 1 | 166 | ||
AB069975.1 | - | 1 | 166 | |||
AK027643.1 | - | 1 | 168 | |||
AK300208.1 | - | 1 | 209 | |||
AL834398.1 | - | 1 | 196 | |||
BC016842.1 | - | 1 | 287 | 3902704 |